Renromsservietter, Bemcot® M-3
Leverandør: CONTEC
M-3
CONTM-3EA 14450 NOK
CONTM-3
Renromsservietter, Bemcot® M-3
Papirservietter Renromsservietter Tørkepapir, renrom
100% rayon non woven wipes for cleaning high-temperature process equipment.
  • 100% rayon, so extremely absorbent
  • Soft and low linting
  • Heat resistant: Ideal for cleaning high-temperature process equipment
  • Used extensively in the compact disk and magnetic tape industries
  • Wipe is quarter folded into a 127×127 mm which is sized to fit the hand and promotes proper wiping protocol
Soft, low linting and heat resistant these wipes are designed for cleaning high temperature process equipment, diffusion furnaces and sensitive electronic equipment in cleanroomMore Product Information