Single use microfibre products, MicroGenesis™
Leverandør: CONTEC
MGMP0001
CONTMGMP0001EA 5525 NOK
CONTMGMP0001
Single use microfibre products, MicroGenesis™
Papirservietter Rengjøringssett og svamper
These single use microfibre pads feature synthetic construction for the effective removal of both gross contamination and fine particles. Conveniently designed to provide a new pad for each new use, they are compatible with most existing hardware.
  • Compatible with common disinfectants like quats, peroxides, and bleach
  • Simplifies quality control, inventory, and space management, all without cross contamination risk
  • Provides efficient and effective removal of surface contamination
  • MicroGenesis™ floor pads and MicroGenesis™ hygienic backer plate can be used