MicroGenesis Mop
Leverandør: CONTEC
MGMP0009
CONTMGMP0009EA 3675 NOK
CONTMGMP0009
MicroGenesis Mop
Mopper
Single-use microfiber mop pads.
  • Synthetic construction is compatible with common disinfectants such as quats, peroxides and bleach
  • New pad for each use simplifies quality control, inventory and space management, all without cross-contamination risk
  • Compatible with most existing hardware
MicroGenesis Mop 100% synthetic, disposable microfiber mop pad effectively removes fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facilities cleaning in ISO Class 8More Product Information