Wipes for delicate tasks, DuPont™ Sontara® Multipurpose Wipe

Leverandør: CONTEC
MP1213BG
CONTMP1213BGEA 4525 NOK
CONTMP1213BG
Wipes for delicate tasks, DuPont™ Sontara® Multipurpose Wipe
Papirservietter Tørkepapir og våtservietter Papirservietter, skånsom
These wipes made from creped spunlaced polyester/cellulose fabric offer outstanding strength and sorbency. Ideal for the removal of thick liquids, greases and dirt.

  • Exceptional performance at an economical price
  • Blue wipes are creped to improve softness
  • White textured wipes scrub, trap and remove contaminants efficiently

Applications: Cleaning in less critical environments, cleaning of glass and smooth surfaces, food processing equipment sanitation and maintenance, and equipment, tool, production line and facility maintenance outside of controlledMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR