Servietter for renrom, Polywipe-C
Leverandør: CONTEC
PC-99 PC-1212
CONTPC-99EA 8400 NOK
CONTPC-99 CONTPC-1212
Servietter for renrom, Polywipe-C
Papirservietter Renromsservietter Tørkepapir, renrom
Polywipe-C Wipes are made from a standardweight 100% knitted polyester that is knife cut to create a soft edge.
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • Low in both particles and extractables to reduce possibility of contamination
  • Good sorbency makes it compatible with many solvents
  • Abrasion and chemical resistant
  • Laundered and packed in an ISO Class 4
The soft edge is ideal for use on surfaces which are vulnerable to scratching. The wipe is chemical resistant and exceptionally low in particles and extractable residueMore Product Information