Cleanroom wipes, pre-saturated, Polynit Heatseal, Prosat®
Leverandør: CONTEC
PSPS0030
CONTPSPS0030EA 18450 NOK
CONTPSPS0030
Cleanroom wipes, pre-saturated, Polynit Heatseal, Prosat®
Papirservietter Renromsservietter Våtservietter, renrom
These wipes are made of 100% knitted polyester pre-saturated with 100% semiconductor grade Isopropyl Alcohol (IPA).
  • No-run interlock knit for good solvent sorbency, abrasion resistance, and chemical resistance
  • Low in particles and extractables
  • Laundered and packaged in an ISO Class 4 cleanroom
Ideal for wiping exterior product surfaces and cleaning the interior surfaces of process tools or other equipment.
Colour: White.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904