PROSAT® Polynit Heatseal Wipes, 10% IPA/90% DIW
Leverandør: CONTEC
PSPS0048
CONTPSPS0048EA 21825 NOK
CONTPSPS0048
PROSAT® Polynit Heatseal Wipes, 10% IPA/90% DIW
Papirservietter Renromsservietter Våtservietter, renrom
These PROSAT® polynit heatseal wipes are presaturated with 10% semi-grade IPA/90% DI water.
  • Reduce particle and fiber generation
  • Abrasion- and chemical-resistant
  • Reduces VOC emissions and solvent use
  • Eliminates the need for in-house filtration
  • Meets USP grade purified water
PROSAT® Polynit heatseal wipes are manufactured from 100% knitted textured polyester with laser-cut sealed edges for very low levels of particles and fibers. Polyester wipes are chemically resistant and exceptionally low in particles and fibers. An interlock knit creates a durable
More Product Information
New Product