PROSAT® ReFIBE Wipes
Leverandør: CONTEC
PSRF1212-BPR PSRF1212IR-BPR PSRF99-BPR PSRF99IR-BPR
CONTPSRF1212-BPREA 12225 NOK
CONTPSRF1212-BPR CONTPSRF1212IR-BPR CONTPSRF99-BPR CONTPSRF99IR-BPR
PROSAT® ReFIBE Wipes
Papirservietter Renromsservietter Våtservietter, renrom
Recycled 100% knitted polyester cleanroom wipes, providing a sustainability story in a single-use world.
  • 100% knitted laundered recycled polyester is abrasion- and chemical-resistant
  • Presaturated with 70% IPA and deionised water
  • Low levels of particles and fibres
  • Soft textured wipes clean surfces without scratching
  • Manufactured from 100% recycled plastic
  • Reduce environmental impact, divert plastic from oceans and landfills help organisations meet sustainability goals
  • Reseable pouch maintains saturation levels throughout use and makes wipe removal easy
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Sustainable Materials
Miljøsertifikat Certificate from recycler