Disinfectant, Contec® Sterile CyChlor
Leverandør: CONTEC
SBC100CC SBT100CC SBC500CC
CONTSBC100CCEA 4200 NOK
CONTSBC100CC CONTSBT100CC CONTSBC500CC
Disinfectant, Contec® Sterile CyChlor
Rengjøringsløsninger Desinfeksjon
Contec® Sterile CyChlor a blend of stabilised hypochlorous acid in purified water and a broad spectrum disinfectant designed for every day use.
  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination
  • Filtered to 0,2 μm and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Suitable for Grade A and B cleanrooms
  • Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of