Cleaning swabs, double-ended, CONSTIX® SC-3
Leverandør: CONTEC
SC-3
CONTSC-3EA 8725 NOK
CONTSC-3
Cleaning swabs, double-ended, CONSTIX® SC-3
Svabere Svabere for rengjøring
SC-3 double-ended round cotton swabs consist of high quality, long staple length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles.
  • Small, round cotton ends
  • High quality cotton fibres provide excellent particle entrapment and retention
  • Tightly wrapped head helps maintain low levels of extractable residue
  • Heads are held to the handles using a methyl-cellulose binder
The high quality cotton fibres provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-More Product Information