Cleaning swabs, sealed foam, CONSTIX® SF-1
Leverandør: CONTEC
SF-1
CONTSF-1EA 4100 NOK
CONTSF-1
Cleaning swabs, sealed foam, CONSTIX® SF-1
Svabere Svabere for rengjøring
The SF-1 sealed foam swab consists of polyurethane foam bonded to a PP handle without the use of adhesives or binders.
  • Polyurethane foam head provides good particle entrapment and removal
  • Large head is ideal for cleaning and applying coating
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal.
Though compatible with many common solvents, the SF-12 is not recommended for use with ketones such as acetone or MEK.
Forpakning: Swabs are packed 500 per case.