Cleaning swabs, sealed foam, CONSTIX® SF-11
Leverandør: CONTEC
SF-12
CONTSF-12EA 1800 NOK
CONTSF-12
Cleaning swabs, sealed foam, CONSTIX® SF-11
Svabere Svabere for rengjøring
The SF-12 sealed foam swab consists of polyurethane foam bonded to a PP handle without the use of adhesives or binders.
  • Small head size enables good access to small features and difficult to reach surfaces
  • Polyurethane open-cell foam head provides good particle entrapment and removal
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery or removal. The SF-12 swab is excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-12 isMore Product Information