Cleaning swabs, sealed foam, CONSTIX® SF-19
Leverandør: CONTEC
SF-19
CONTSF-19EA 3425 NOK
CONTSF-19
Cleaning swabs, sealed foam, CONSTIX® SF-19
Svabere Svabere for rengjøring
The SF-19 sealed foam swab consists of a 100 PPI sealed closed-cell polyurethane foam head formed over the cotton bud of a 6” applicator with wooden handle.
  • Soft polyurethane foam head provides good particle entrapment and removal
  • Wooden handle is solvent resistant and stiff enough to allow vigorous swabbing
  • No adhesives are used
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery and removal. The SF-19 swab is very low in particles and fibres, as well suited toMore Product Information