Cleaning swabs, sealed foam, CONSTIX® SF-3
Leverandør: CONTEC
SF-3
CONTSF-3EA 2775 NOK
CONTSF-3
Cleaning swabs, sealed foam, CONSTIX® SF-3
Svabere Svabere for rengjøring
The SF-3 swab consists of 100 pore/inch sealed closed cell foam head with a PP handle.
  • Swab head is thermally sealed and bonded to the handle without adhesives or binders
  • Large head is ideal for cleaning and applying coatings
The polyurethane foam head provides good particle entrapment and removal.
The SF-3 swab is compatible with many solvents, such as isopropanol and ethanol, but is not recommended for use with ketones, such as acetone or MEK.
Forpakning: Swabs are packed 500 per case.
NB: *Produktet/produktene på denne siden er utenforMore Product Information