Cleaning swabs, sealed foam, CONSTIX® SF-6
Leverandør: CONTEC
SF-6
CONTSF-6EA 5350 NOK
CONTSF-6
Cleaning swabs, sealed foam, CONSTIX® SF-6
Svabere Svabere for rengjøring
The SF-6 sealed foam swab consists of a 100 PPI polyurethane foam adhesive bonded to a flexible nylon tip. The tip is mechanically secured within a tubular PP handle.
  • Flexible nylon tip allows for a softer application
  • Polyurethane foam head provides good particle entrapment for removal and solvent compatibility
The absorbant foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal.
Though compatible with many common solvents, the SF-6 is not recommended for use with ketonesMore Product Information