Cleaning swabs, sealed, CONSTIX® SP-10
Leverandør: CONTEC
SP-10
CONTSP-10EA 2275 NOK
CONTSP-10
Cleaning swabs, sealed, CONSTIX® SP-10
Svabere Svabere for rengjøring
CONSTIX® SP-10 sealed polyester swab consists of 100% knitted polyester fabric that is thermally bonded to a flex-tip PP handle.
  • Double-layer, sealed, knitted polyester swab
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Polyester head is compatible with most common solvents, such as alcohols and ketones
The swab head is made of cleanroom laundered; double-layer knitted polyester material. The polyester head is thermally sealed and attached, also constructed without adhesives or binders. The polypropylene handle has a thin,More Product Information