Cleaning swabs, sealed, CONSTIX® SP-6
Leverandør: CONTEC
SP-6
CONTSP-6EA 4250 NOK
CONTSP-6
Cleaning swabs, sealed, CONSTIX® SP-6
Svabere Svabere for rengjøring
The SP-6 sealed polyester swab consists of 100% knit polyester fabric that is thermally bonded to a flex-tip PP handle.
  • Flexible tip provides a softer application on surfaces
  • 100% polyester head provides enhanced sorbency, particle entrapment, and removal
  • Head is compatible with most common solvents, such as alcohols and ketones
  • Durable and absorbent
  • Extremely low particles and residue levels
The swab head is made of cleanroom laundered; double-layer knitted polyester. The polyester head is constructed without adhesives or binders.
Forpakning: 500More Product Information