QuickTask™ Microfibre Mop Heads
Leverandør: CONTEC
TASK0800 TASK0850
CONTTASK0800EA 3725 NOK
CONTTASK0800 CONTTASK0850
QuickTask™ Microfibre Mop Heads
Mopper
Single-use, 100% polyester single-loop microfibre flat mop head.
  • 100% single-loop microfibre mop heads are compatible with a wide range of disinfectants and detergents
  • Low levels of particles and fibres
  • High absorbency saves times and reduces the number of visits back to a bucket system
  • Approximately 22 m² coverage per mop
  • Ideal for presaturation systems
  • Available sterile, which is suitable for use in Grade B cleanrooms
  • Double-bagged packaging facilitates transfer into cleanrooms and critical environments
  • Suitable for hands-free mop application (with