4N6FLOQSwabs® Forensic DNA Collection for Human Identification
Leverandør: Copan
4520CS01 4508C 4504C 4520CA 40U022D
COPA4520CS01EA 18875 NOK
COPA4520CS01 COPA4508C COPA4504C COPA4520CA COPA40U022D
4N6FLOQSwabs® Forensic DNA Collection for Human Identification
Svabere Svabere for prøvetaking
DNA collection for human identification. The active drying system absorbs water molecules, preventing microbial growth and guaranteeing DNA stability for 12+ months at RT and –80 °C.
  • Application: High performance sample collection 
  • FLOQ® technology: this Swab is empowered by the FLOQ® technology for superior sample adsorption and release
  • 20 mm breaking point: the short length breaking point and the 2-1,5 ml vials compatible tip facilitate the laboratory workflow
  • Gold standard for forensic DNA evidence collection: 4N6FLOQSwabs® w/Active drying