Multicoated microplates
Leverandør: Corning
3829
CORN3829EA 21200 NOK
CORN3829
Multicoated microplates
Mikroplater Celledyrkningsplater
The Corning® multicoated microplate provides users access to six different surface treatments on a single plate.
This microplate is useful when determining the correct surface for assay requirements. Single surface microplates can then be used for the full screen or experiment. These multicoated plates are currently available in 96-well, 384-well, and 1536-well formats in black with clear bottom.