Erlenmeyer Flask, wide neck, graduated, Corning®
Leverandør: Corning
431510
CORN431510EA 8500 NOK
CORN431510
Erlenmeyer Flask, wide neck, graduated, Corning®
Kolber Erlenmeyerkolber
This flask is made from optically clear polycarbonate and features an orange PP cap.
  • With 1/8" dip tube, 0,2 μm vent, male luer lock
  • Sterilized by gamma radiation and nonpyrogenic
  • Flat bottom design
  • Wide mouth neck style
  • Graduated (graduation interval: 50 ml)
The preassembled transfer cap has two ports. One port ends in a 0.2 µm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in either a male Luer lock or a male polycarbonate quick connect. The dip tube reaches all the way to the bottom of the flask for easyMore Product Information