Ultra-low attachment 25 cm² rectangular canted neck cell culture flask with vent cap, Corning®
Leverandør: Corning
4616
CORN4616EA 3850 NOK
CORN4616
Ultra-low attachment 25 cm² rectangular canted neck cell culture flask with vent cap, Corning®
Mikrobiologiske medier Media, preparerte for mikrobiologi
Features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation.
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilised by gamma irradiation
  • Nonpyrogenic
The Corning vessels feature a biologically inert ultra-low attachment surface that minimizes cell attachment and promotes the formation of 3D multicellular spheroids employed in cancer research, stem cell biology, and drug screening.
Minimises cell attachment, protein absorption, and enzyme activation.