1536-well spheroid microplates, Corning®
Leverandør: Corning
4637
CORN4637EA 7700 NOK
CORN4637
1536-well spheroid microplates, Corning®
Mikroplater Celledyrkningsplater
With their proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • 536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion