Serological pipettes, Corning®
Leverandør: Corning
7374
CORN7374EA 0 NOK
CORN7374
Serological pipettes, Corning®
Pipetter Serologiske pipetter
Sterile serological pipettes made of PS.
Forpakning: Triple bagged.
NB: These pipettes are "Made To Order" products. The minimum order quantity is 40 cases. Please search for Stripette® pipettes when looking for smaller order quantities.