Magnetic stirring bars, double cross-head, PTFE
Leverandør: COWIE
001.1135
COWI001.1135EA 200 NOK
COWI001.1135
Magnetic stirring bars, double cross-head, PTFE
Omrørere Magnetrørerstaver
Double cross-head has a greater turbulance when compared to single cross-head.