D-rings, DBI-SALA, 3M™
Leverandør: CAPITAL SAFETY
1500003
CPSD1500003EA 0 NOK
CPSD1500003
D-rings, DBI-SALA, 3M™
Fallvern Fallvern
D-rings can be attached to virtually any tool under 2,3 kg (5 lbs.) or 0,9 kg (2 lbs.) by using Quick-Wrap Tape.
  • Creates an attachment point in less than a minute
  • Dual D-rings allow for 100% tool tie-off during tool transfers