D-Ring attachment with cord, DBI-SALA, 3M™
Leverandør: CAPITAL SAFETY
1500009
CPSD1500009EA 0 NOK
CPSD1500009
D-Ring attachment with cord, DBI-SALA, 3M™
Fallvern Fallvern
D-Ring cord attachment point, 2,3 kg (5 lbs.) capacity.
  • Adjusts to virtually any wrist size with hook and loop system
  • Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
  • Elastic band helps ensure a comfortable fit
  • Attached trigger can be used to quickly work with many hand tools
The DBI SALA® D-ring cord easily cinches to tools weighing under 2,3 kg creating an instant attachment point for many tools.
Designed for use in general industrial, oil and gas, mining,More Product Information