Adjustable wristbands, DBI-SALA®
Leverandør: CAPITAL SAFETY
1500085 1500087
CPSD1500085EA 0 NOK
CPSD1500085 CPSD1500087
Adjustable wristbands, DBI-SALA®
Bånd mot elektrostatisk utladning Håndleddbånd mot elektrostatisk utladning
Adjustable wristbands are available with retractor and elastic strap with hook and loop, or with cord and elastic strap with hook and loop.
  • Adjusts to virtually any wrist size with hook and loop system
  • Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
  • Elastic band helps ensure a comfortable fit
  • Attached trigger can be used to quickly work with many hand tools
The DBI-SALA® Adjustable wristband with retractor is constructed out of elastic webbing that can beMore Product Information