Self-retracting lifeline, Protecta® Rebel™, 3M™
Leverandør: CAPITAL SAFETY
3100437
CPSD3100437EA 0 NOK
CPSD3100437
Self-retracting lifeline, Protecta® Rebel™, 3M™
Fallvern Falltau
The robust Rebel SRL is made from polyester web with zinc plated snap hook 19 mm opening.
  • Swiveling anchorage loop and carabiner
  • 25 mm polyester web lifeline
  • Self-locking snap hook on lifeline
  • Compact, lightweight design
  • Tough and impact resistant stackable thermoplastic housing
  • Quick activating speed sensing brake system
  • High capacity external energy absorber and impact indicator
Length: 6,1 m
Capacity: 141 kg
Rebel cable blocks meets the latest CE sharp edge certifcation when used with an extension sling.
Sertifiseringer: EN 360