Aluminium tripod, DBI-SALA, 3M™
Leverandør: CAPITAL SAFETY
8563158 8563159
CPSD8563158EA 0 NOK
CPSD8563158 CPSD8563159
Aluminium tripod, DBI-SALA, 3M™
Fallvern Fallvern
Tripod with adjustable locking legs, safety chains, top pulley and winch bracket.
  • Available in two heights
Weight capacity: 205 kg
Sertifiseringer: EN 795 Class-B
Bestillingsinformasjon: Winch and SRL are not included.
Bestill nå
SPESIFIKASJONER
Modell UCT-300 UCT-1000