D-ring anchorage, Protecta®, 3M™
Leverandør: CAPITAL SAFETY
AM211
CPSDAM211EA 0 NOK
CPSDAM211
D-ring anchorage, Protecta®, 3M™
Fallvern Fallvern
Protecta® anchorage D-ring made from 316 stainless steel; 12 mm fixing.
  • Can be used with three load directions
Sertifiseringer: EN 795 Class A
Bestillingsinformasjon: Supplied without fixing.