Supelco® TraceCERT® Organic Reference Standard, Tetramethrin

Leverandør: Merck

CRM03054-100MG
CRM03054-100MGEA 1275 NOK
CRM03054-100MG
Supelco® TraceCERT® Organic Reference Standard, Tetramethrin
Standarder Organiske standarder

Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


NB: P201 - P202 - P273 - P301 + P312 - P304 + P340 + P311 - P308 + P311

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 7696-12-0
MDL No. MFCD00084635
Molekylvekt 331,41
Hardhetsklasse beskrivelse H302 - H331 - H351 - H371 - H410
Fraktforhold WICE
Lagringsforhold –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR