Supelco® TraceCERT® Organic Reference Standard, Diethofencarb
Leverandør: Merck
CRM04342-25MG
CRM04342-25MGEA 700 NOK
CRM04342-25MG
Supelco® TraceCERT® Organic Reference Standard, Diethofencarb
Standarder Organiske standarder
Diethofencarb may be used as certified reference material for the following:
  • To develop a dispersive liquid-liquid microextraction method based on the solidification of a floating organic droplet (DLLME-SFO) for simultaneous analysis of diethofencarb and pyrimethanil in apple pulp and peel using liquid chromatography with diode-array detection (HPLC-DAD)
  • To investigate the sensitivity profile of Botrytis cinerea field isolates to zoxamide and evaluate the resistance mechanism involved in cross-resistance relationships between benzamides benzimidazoles and N-phenylcarbamates
  • To develop an ultrasound-assisted surfactant-enhanced emulsification microextraction method for the analysis of diethofencarb and pyrimethanil fungicides in water and fruit juice samples using high performance liquid chromatography-diode array detector/electrospray ionisation mass spectrometry (HPLC-DAD/ESI-MS)
  • Preparation of 3D nanotubes assembly from MoS₂ monolayers (3D MoS₂NTs) for rapid and selective screening of diethofencarb
  • Development of a dispersive liquid-liquid microextraction method for the determination of diethofencarb and pyrimethanil in aqueous samples
NB: P264 - P280 - P305 + P351 + P338 - P337 + P313