Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb-sulfone

Leverandør: Merck

CRM05139-10MG
CRM05139-10MGEA 2175 NOK
CRM05139-10MG
Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb-sulfone
Standarder Organiske standarder

Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


NB: P264 - P270 - P301 + P312 - P501

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 53380-23-7
MDL No. MFCD01311808
Molekylvekt 257,31
Hardhetsklasse beskrivelse H302
Fraktforhold WICE
Lagringsforhold 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR