Supelco® TraceCERT® Organic Reference Standard, Fenobucarb
Leverandør: Merck
CRM05583-100MG
CRM05583-100MGEA 1425 NOK
CRM05583-100MG
Supelco® TraceCERT® Organic Reference Standard, Fenobucarb
Standarder Organiske standarder
Fenobucarb may be used as reference material for the following:
  • Method development and validation for simultaneous determination of fenobucarb enantiomers in vegetables crops soils and water by UPLC-MS/MS using a Daicel IG-3 chiral column
  • Method development for determination of fenobucarb residues in vegetable samples using screen-printed carbon electrode modified with graphene nanoribbons-ionic liquid-cobalt phthalocyanine composites based on flow injection analysis (FIA)
  • Quantification of fenobucarb residues in animal (porcine muscle egg and whole milk) and aquatic (eel flatfish and shrimp) food products using QuEChERS extraction and liquid chromatography-electrospray ionisation tandem mass spectrometry (LC-ESI/MS-MS)
  • Development of an electrochemical screening assay for the detection of 4 phenyl carbamates in grains samples and comparing the data with UHPLC-MS/MS
  • Quantitative analysis of fenobucarb residues in beef muscles using QuEChERS method and liquid chromatography-tandem mass spectrometry
NB: P264 - P270 - P273 - P301 + P312 - P391 - P501