Supelco® TraceCERT® Organic Reference Standard, Acequinocyl
Leverandør: Merck
CRM52422-50MG
CRM52422-50MGEA 1700 NOK
CRM52422-50MG
Supelco® TraceCERT® Organic Reference Standard, Acequinocyl
Standarder Organiske standarder
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
NB: P260 - P264 - P273 - P280 - P302 + P352 - P308 + P311