Picric acid 13 g/l saturated solution in water

Leverandør: CLIN TECH

Synonymer: 2,4,6-Trinitrophenol, TNP

640760
CTEH640760EA 0 NOK
CTEH640760
Picric acid 13 g/l saturated solution in water
Picric acid
Formel: C₆H₃N₃O₇
MDL nummer: MFCD00007102
CAS-nummer: 88-89-1
EINECS: 201-865-9
Merck Indeks: 13,07492

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR