Fungal DNA kits, E.Z.N.A.®

Leverandør: OMEGA BIO-TEK

D3390-02
D3390-02EA 0 NOK
D3390-02
Fungal DNA kits, E.Z.N.A.®
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit

The E.Z.N.A.® Fungal DNA kits allow for the rapid and reliable isolation of high quality total cellular DNA from a wide variety of fungal species without the need for organic extraction.

Bestill nå

SPESIFIKASJONER

Beskrivelse E.Z.N.A.® Fungal DNA Mini Kit
Environmentally Preferable
Applikasjon
Target molecule DNA
Refine target molecule Total DNA
Sample type Yeast/Fungi
Method / Format Column (silica-based)
Prep size Mini
Prøvestørrelse 100 mg wet or 30 mg dry tissue

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR