E.Z.N.A.® Blood DNA Kit
Leverandør: OMEGA BIO-TEK
D3392-02
D3392-02EA 4750 NOK
D3392-02
E.Z.N.A.® Blood DNA Kit
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
The E.Z.N.A.® blood DNA kit provides rapid total DNA isolation from up to 250 µl of fresh or frozen anticoagulated whole blood. The E.Z.N.A.® blood DNA kit can also be used for the preparation of genomic DNA from buffy coat, serum, plasma, bone marrow, lymphocytes, platelets, and body fluids. This kit allows for simultaneous processing of single or multiple samples in less than 30 minutes. Phenol/chloroform extractions, and time-consuming steps such as precipitation with isopropanol or ethanol have been eliminated. DNA purified with the E.Z.N.A.® blood DNA method is ready for applications such as PCR, southern blotting, or restriction enzyme digestion.
  • Isolation of DNA in less than 30 minutes
  • Optimised buffer system ensures reproducible results
  • No organic extractions
  • Purified DNA suitable for most downstream applications