E.Z.N.A.® SQ DNA-kit for blod
Leverandør: OMEGA BIO-TEK
D5032-03
D5032-03EA 0 NOK
D5032-03
E.Z.N.A.® SQ DNA-kit for blod
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
The E.Z.N.A.® SQ blood DNA kit provides a reliable method for the isolation of high molecular weight genomic DNA from fresh, frozen, or anti-coagulated whole blood. The method can also be used for preparation of genomic DNA from buffy coat, bone marrow or cultured cells. There is no need for phenol/chloroform extractions, and time consuming steps such as CsCl gradient ultracentrifugation are eliminated. DNA purified using the E.Z.N.A.® SQ blood DNA method is ready for downstream applications such as PCR, southern blotting and restriction enzyme digestion.
  • Isolering av høy kvalitet, genomisk DNA med høy molekylær vekt
  • Fleksibilitet i utgangsmateriale med DNA-rensing fra 0,1 til 20 ml fullblod
  • Behandle én eller flere prøver på mindre enn 90 minutter.