2'-Deoxycytidine hydrochloride, Supelco®

Leverandør: Merck

Synonymer: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

D8006-10G D8006-1G
D8006-10GEA 6125 NOK
D8006-10G D8006-1G
2'-Deoxycytidine hydrochloride, Supelco®
2'-Deoxycytidine hydrochloride
Formel: C₉H₁₄ClN₃O₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012840
CAS-nummer: 3992-42-5
EINECS: 223-639-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d99a5fd51698aeff205afe10ad652e77632eb8&vid=e6d99a5fd51698aeff205afe10ad652e77632eb8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e30612856c3bd857017a2fbf0c2da16524d77a36&vid=e30612856c3bd857017a2fbf0c2da16524d77a36&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR