Goggles, Vistamax VNC21
Leverandør: HONEYWELL SAFETY
1002759
DALL1002759EA 270 NOK
DALL1002759
Goggles, Vistamax VNC21
Vernebriller /Goggles
High performance anti-fog goggles, suitable for most very humid environments.
  • No vent frame gives excellent protection againsts large and fine dust particles as well as droplets of liquids
  • Unique anti-fog twin lens system: Front acetate lens for chemical resistance, rear polycarbonate lens for impact resistance
  • Thermal bridge with aluminimum condenser
Designed for petrochemical, industrial or laboratory, administration or foundry.
Sertifiseringer: ISO 9001/2000