Filter Bags for Homogeniser
Leverandør: DELTALAB
15005
DELT15005EA 14225 NOK
DELT15005
Filter Bags for Homogeniser
Poser Homogenisator-poser
These are sterile bags having a full size filter dividing them into two identical pouches, in which sample is inserted into one compartment. After sample process, solid particles remain in their original compartment while liquid will be filtered into the other compartment, thus allowing a convenient liquid pipetting.
  • Beta irradiation
  • Graduated for marking