Weighing boats
Leverandør: DELTALAB
1911103
DELT1911103EA 1325 NOK
DELT1911103
Weighing boats
Skip Veieskip
PS, square, white.
  • Resistant to weak acids, water based solutions and alcohol
  • Can be folded for easy pouring
  • Suitable for microwave