Liquid Amies, Sterile, Transporter Swab
Leverandør: DELTALAB
300284.SE
DELT300284.SEEA 13150 NOK
DELT300284.SE
Liquid Amies, Sterile, Transporter Swab
Svabere Svabere for transport
This is a variation of the traditional Amies media swab where the media is presented on liquid form without agar.
  • Peel-pack dimensions: 38×210 mm
Media is supplied in its liquid form, in a sponge, suitable for direct extensions on slides.
Swabs are sterilised by radiation.
Suitable for conservation of pathogens such as:Haemophilus sp., Corynebacterium sp., Streptococcus pyogenes, Streptococcus pneumoniae, Shigella flexneri, Salmonella typhi, Brucella abortus, Staphylococcus epidermidis, Escherichia coli.
Sertifiseringer: CE MDD Class IIa. CEMore Product Information