Strips of 12 PCR tubes and cap strips
Leverandør: DELTALAB
4095.4N
DELT4095.4NEA 6200 NOK
DELT4095.4N
Strips of 12 PCR tubes and cap strips
Rør PCR-rør
Tubes made of PP.
  • RNAse, DNAse and pyrogen free
  • Excellent chemical resistance
  • Excellent chemical resistance
Cap and tube strips allow easy handling and inventory of tubes and caps. It also prevent cross-contamination from tube to tube.