Container, wide neck, cylindrical, with screw cap
Leverandør: DELTALAB
44430000
DELT44430000EA 0 NOK
DELT44430000
Container, wide neck, cylindrical, with screw cap
Prøvecontainere Beholdere, generelt bruk
HDPE, translucent, with star-shaped cap.
  • Good chemical resistance
  • Good temperature resistance
Tagging points on neck and cap for a tamper proof closure or for labelling.
Leveringsinformasjon: Containers are supplied non capped.