Tubing, PVC, with reinforcing webbing
Leverandør: DEUTSCH NEUMANN
3510915 3511320 3510814 3511927 3512534 3511221 3511624 3513242 3510612 3511016 3511219 3513848 3510410 3510511 3500810A
228-1727EA 98 NOK
228-1727 228-1729 DENE3510814 DENE3511927 DENE3512534 DENE3511221 DENE3511624 DENE3513242 DENE3510612 DENE3511016 DENE3511219 DENE3513848 DENE3510410 DENE3510511 228-3843
Tubing, PVC, with reinforcing webbing
Slanger Overføringsslange
PVC, transparent.
The clear implementation and the wide-mashed inlay enables a flawless observation of the fluid. This tubing features a good resistance against chemicals, aging, oil, and fuel.
Temperature resistance: up to 60 °C
Hardness (Shore durometer A): 75