Rubber protection mats
Leverandør: DEUTSCH NEUMANN
5202004 5203003 5202001 5202002 5203001 5203002 5203004 5202015 5203015 5202003
DENE5202004EA 4825 NOK
DENE5202004 DENE5203003 DENE5202001 DENE5202002 DENE5203001 DENE5203002 DENE5203004 DENE5202015 DENE5203015 DENE5202003
Rubber protection mats
Benkepapir Benkepapir
Black protection sheets made from fluoroelastomer rubber (FPM). Durometer 75 Shore A.
  • For temperature ranges from −10 to over +200 °C
  • Good resistance to ozone and weathering
  • Benzine and mineral oil resistant