Metal-In shield bags, PCL100 Clean series
Leverandør: DESCO
PCL1001115
DESCPCL1001115EA 565 NOK
DESCPCL1001115
Metal-In shield bags, PCL100 Clean series
Poser Oppbevaringsposer
Metal-In static shielding bags are manufactured from a polyester, metal, polyethylene laminate. ESD safe packaging designed for use in cleanroom areas where surface particulate is of concern.
  • Embossed with ESD protective symbol and lot code information
  • The polyester dielectric in concert with the metal layer provides discharge shielding
  • Bag is free of amines, silicones and heavy metals
  • Bags are heat sealable
Metal-In film laminate 0,07 mm protects ESD sensitive contents from electrostatic fields and electrostatic discharges (ESD). Dissipative innerMore Product Information